Learn More
Cellvis 29 mm Glass bottom dish with 20 mm micro-well #1.5 cover glass, 100/case
Catalog No. NC0636070
Encompass
Suitable for long term tissue culture.Manufactured in a class 100,000 clean room.Dish made from virgin polystyrene, tissue culture treated.German cover glass of superior optical quality.A USP class VI adhesive is used to assemble the cover glass and the dish.Packed in easy to open peelable bag.Sterilized by Gamma radiation.
Catalog No. NC0636070 Supplier Cellvis Supplier No. D29201.5N
Suitable for long term tissue culture.Manufactured in a class 100,000 clean room.Dish made from virgin polystyrene, tissue culture treated.German cover glass of superior optical quality.A USP class VI adhesive is used to assemble the cover glass and the dish.Packed in easy to open peelable bag.Sterilized by Gamma radiation.