Learn More
IDEXX REFERENCE LABORATORIES 290 ML VESSEL W/N/100CS
Catalog No. NC0551498
Encompass
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals. High clarity and nonfluorescing polystyrene make it easy to see color and fluorescent end points ; Gamma sterilization eliminates false positives due to contamination (certificate of sterility can be kept for audit purposes) ; 100 mL line around entire perimeter eliminates need for sterile graduated cylinder (meets U.S. EPA guidelines for +/- 2.5% accuracy) ; Recyclable Type 6 polystyrene minimizes environmental impact ; Sodium thiosulfate option neutralizes at least 15 mg/Liter of chlorine (applies to 120ml and 150ml vessels only) ; Multiple sizes of 120-, 150- and 290-mL vessels provide greater or added flexib
This item is not returnable. View return policy
Catalog No. NC0551498 Supplier IDEXX REFERENCE LABORATORIES Supplier No. WV290SBST100
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals. High clarity and nonfluorescing polystyrene make it easy to see color and fluorescent end points ; Gamma sterilization eliminates false positives due to contamination (certificate of sterility can be kept for audit purposes) ; 100 mL line around entire perimeter eliminates need for sterile graduated cylinder (meets U.S. EPA guidelines for +/- 2.5% accuracy) ; Recyclable Type 6 polystyrene minimizes environmental impact ; Sodium thiosulfate option neutralizes at least 15 mg/Liter of chlorine (applies to 120ml and 150ml vessels only) ; Multiple sizes of 120-, 150- and 290-mL vessels provide greater or added flexib