Learn More
Ace Glass, Inc. 290mm B quartz immersion well, Tube connection
SDP
Catalog No. 50975520
Encompass
290mm B double-walled quartz immersion well with inlet and outlet tubes for cooling. Inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well has 60/40 inner joint. O.D. of outer jacket is 53mm max. Length above joint is 76mm.
Catalog No. 50-975-520 Supplier Ace Glass, Inc. Supplier No. 785427
290mm B double-walled quartz immersion well with inlet and outlet tubes for cooling. Inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well has 60/40 inner joint. O.D. of outer jacket is 53mm max. Length above joint is 76mm.