Learn More
Medchemexpress LLC HY-107994 1g Medchemexpress, Aminooxyacetic acid hemihydrochloride Carboxymethoxylamine Hemihydrochloride CAS:2921-14-4 Purity:98%
SDP
Catalog No. 501637373
Encompass_Preferred
Medchemexpress,HY-107994 1g Aminooxyacetic acid hemihydrochloride Carboxymethoxylamine Hemihydrochloride CAS:2921-14-4 Formula:NH2OCH2COOH · 0.5HCl Aminooxyacetic acid hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the GABA degradating enzyme GABA-T. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7373 Supplier Medchemexpress LLC Supplier No. HY1079941G
Medchemexpress,HY-107994 1g Aminooxyacetic acid hemihydrochloride Carboxymethoxylamine Hemihydrochloride CAS:2921-14-4 Formula:NH2OCH2COOH · 0.5HCl Aminooxyacetic acid hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the GABA degradating enzyme GABA-T. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.