Learn More
Invivogen 293-DUAL HSTING-R232 CELLS 3-7
SDP
Catalog No. NC2593982
Encompass
Dual IRF and IFN-β reporter 293 cells expressing R232 isoform of human STING, 3-7 x 10e6 cells
Catalog No. NC2593982 Supplier Invivogen Supplier No. 293DR232
Add to Cart
Add to Cart
Dual IRF and IFN-β reporter 293 cells expressing R232 isoform of human STING, 3-7 x 10e6 cells