Learn More
Cell Biolabs Inc HEK 293/GFP Cell Line
Catalog No. 50672586
Encompass_Preferred
Cell Biolabs' 293/GFP Cell Line stably expresses GFP and blasticidin-resistant genes. Both GFP and blasticidin-resistant genes were introduced into parental 293 cells using lentivirus. This cell line otherwise exhibits the same characteristics of the parental 293 cell line.
Catalog No. 50-672-586 Supplier Cell Biolabs Inc Supplier No. AKR200
Cell Biolabs' 293/GFP Cell Line stably expresses GFP and blasticidin-resistant genes. Both GFP and blasticidin-resistant genes were introduced into parental 293 cells using lentivirus. This cell line otherwise exhibits the same characteristics of the parental 293 cell line.