Learn More
System Biosciences LLC 293TN PRODUCER CELL LINE293TN Producer Cell Line = 2 x 10^6 Cells
SDP
Catalog No. NC1116896
Encompass_Preferred
Delivering high titer lentivirus prepsOptimized for effective, high-titer production of pseudoviral particles, SBI’s 293TN Producer Cell Line has been transformed with the SV40 large T antigen to promote robust growth and features the neomycin resistance marker for stable propagation. The 293TN packaging cell line is shipped at low passage (P2).
Catalog No. NC1116896 Supplier System Biosciences LLC Supplier No. LV900A1
Delivering high titer lentivirus prepsOptimized for effective, high-titer production of pseudoviral particles, SBI’s 293TN Producer Cell Line has been transformed with the SV40 large T antigen to promote robust growth and features the neomycin resistance marker for stable propagation. The 293TN packaging cell line is shipped at low passage (P2).