Learn More
ATCC 293TT; Kidney Epithelium; Human (Homo sapiens)
Supplier:  ATCC CRL3467
Encompass
293TT is a cell line exhibiting epithelial morphology that was isolated from a human kidney. It has applications for production of papillomavirus and polyomavirus-based reporter vectors, as well as propagation of torque teno viruses, BK and JC polyomavirus primary isolates.
Catalog No. 50-238-2052