Learn More
Sino Biological Recombinant Human EGFR Protein (Isoform Viii, hFc Tag)
Supplier:  Sino Biological 29662H02B50
Encompass_Preferred
A DNA sequence encoding the human EGFR (NP_001333870.1) (Met1-Lys375) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0033