Learn More
Sino Biological Recombinant Human PCSK9 Protein (mFc Tag)
Supplier:  Sino Biological 29698H05H50
Encompass_Preferred
A DNA sequence encoding the human PCSK9 ( NP_777596.2) (Met1-Gln692) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-162-0040