Learn More
Waters Corp Waters Corporation Analytical Flowcell, TaperSlit, 10 mm Pathlength, 8.4 μL Volume
Catalog No. 50791674 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This analytical flowcell features a TaperSlit design, a 10 mm pathlength, and an 8.4 μL volume, making it suitable for use with Platinum HPLC Detectors.
  • Features a TaperSlit design
  • Provides a 10 mm pathlength
  • Offers an 8.4 μL volume
  • Compatible with Platinum HPLC Detectors
Catalog No. 50-791-674 Supplier Waters Corp Supplier No. 205000399
This analytical flowcell features a TaperSlit design, a 10 mm pathlength, and an 8.4 μL volume, making it suitable for use with Platinum HPLC Detectors.
  • Features a TaperSlit design
  • Provides a 10 mm pathlength
  • Offers an 8.4 μL volume
  • Compatible with Platinum HPLC Detectors