Learn More
Waters Corp Waters Flowcell Rebuild Kit, for 2998 Analytical Flowcell, HPLC and UPLC
Catalog No. 50792912 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains the lenses and gaskets necessary to rebuild a 2998 analytical flow cell.
  • Designed for analytical flow cell rebuilds.
  • Compatible with HPLC and UPLC systems.
  • Suitable for use with HPLC detectors, UPLC detectors, and ACQUITY Arc instruments.
  • Contains all necessary components for rebuilding a 2998 PDA detector.
  • Replacement as required to maintain instrument performance.
  • Supplied as 1 unit per package.
Catalog No. 50-792-912 Supplier Waters Corp Supplier No. 700003798
This rebuild kit contains the lenses and gaskets necessary to rebuild a 2998 analytical flow cell.
  • Designed for analytical flow cell rebuilds.
  • Compatible with HPLC and UPLC systems.
  • Suitable for use with HPLC detectors, UPLC detectors, and ACQUITY Arc instruments.
  • Contains all necessary components for rebuilding a 2998 PDA detector.
  • Replacement as required to maintain instrument performance.
  • Supplied as 1 unit per package.