Learn More
Waters Corp WATERS CORPORATION Semi-preparative Flow Cell Kit, 3 mm Path Length, 10.2 μL Volume
Catalog No. 50791676 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This semi-preparative flow cell kit is designed for optimal performance in various laboratory applications, ensuring accurate and reliable results.
  • 3 mm path length
  • 10.2 μL volume
Catalog No. 50-791-676 Supplier Waters Corp Supplier No. 205000401
This semi-preparative flow cell kit is designed for optimal performance in various laboratory applications, ensuring accurate and reliable results.
  • 3 mm path length
  • 10.2 μL volume