Learn More
Savillex Corporation 29 mm Closure
SDP
Catalog No. NC1612142
Encompass
Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C). Knurled finish on the closure. Leakproof seal.
Catalog No. NC1612142 Supplier Savillex Corporation Supplier No. 60002901
Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C). Knurled finish on the closure. Leakproof seal.