Learn More
Cellvis 29 mm Glass bottom dish with 10 mm micro-well #0 cover glass
Catalog No. NC1831398
Encompass
29mm glass bottom dish, dish size 29mm, well size 10mm, #0 cover glass(0.085-0.115mm). Designed for high resolution imaging such as confocal microscopy. Suitable for long term tissue culture. Manufactured in a class 100,000 clean room. Dish made from virgin polystyrene, tissue culture treated. German cover glass of superior optical quality. A USP class VI adhesive is used to assemble the cover glass and the dish. Packed in easy to open peelable bag. Sterilized by Gamma radiation.
Catalog No. NC1831398 Supplier Cellvis Supplier No. D29100N
29mm glass bottom dish, dish size 29mm, well size 10mm, #0 cover glass(0.085-0.115mm). Designed for high resolution imaging such as confocal microscopy. Suitable for long term tissue culture. Manufactured in a class 100,000 clean room. Dish made from virgin polystyrene, tissue culture treated. German cover glass of superior optical quality. A USP class VI adhesive is used to assemble the cover glass and the dish. Packed in easy to open peelable bag. Sterilized by Gamma radiation.