Learn More
Cellvis 29MM GL BTM DISH COVR GL 100CS
Catalog No. NC2394837
Encompass
Suitable for long term tissue cultureManufactured in a class 100,000 clean roomDish made from virgin polystyrene, tissue culture treated.German cover glass of superior optical qualityA USP class VI adhesive is used to assemble the cover glass and the dish.Packed in easy to open peelable bagSterilized by Gamma radiation.
Catalog No. NC2394837 Supplier Cellvis Supplier No. D29201.5H
Suitable for long term tissue cultureManufactured in a class 100,000 clean roomDish made from virgin polystyrene, tissue culture treated.German cover glass of superior optical qualityA USP class VI adhesive is used to assemble the cover glass and the dish.Packed in easy to open peelable bagSterilized by Gamma radiation.