Learn More
Cayman Chemical 2α-Mannobiose, Purity: ≥95%, CAS: 15548-39-7, 10 mg
SDP
Catalog No. 502985984
Encompass_Preferred
A disaccharide; composed of two mannose molecules linked by a 1-2 glycosidic bond; has been immobilized on monolithic capillary columns for the affinity purification of mannose binding proteins
Catalog No. 50-298-5984 Supplier Cayman Chemical Supplier No. 2184810 MG
A disaccharide; composed of two mannose molecules linked by a 1-2 glycosidic bond; has been immobilized on monolithic capillary columns for the affinity purification of mannose binding proteins