Learn More
Cayman Chemical 2AmIno2deoxyadenosIn5 2.5mg
SDP
Catalog No. 502915960
Encompass_Preferred
An amino-substituted derivative of ATP; has been used to determine the substrate specificity of modified DNA polymerases
Catalog No. 50-291-5960 Supplier Cayman Chemical Supplier No. 383622.5 MG
An amino-substituted derivative of ATP; has been used to determine the substrate specificity of modified DNA polymerases