Learn More
Cayman Chemical 2AmIno3 4dImethyl3HImIda 10mg
SDP
Catalog No. 502991857
Encompass_Preferred
A mutagenic heterocyclic amine; mutagenic to S. typhimurium in the Ames test; induces tumor formation in the Zymbal gland, oral cavity, and colon in male and female rats, the skin of male rats, and the mammary gland in female rats at 0.03% in the diet
Catalog No. 50-299-1857 Supplier Cayman Chemical Supplier No. 3150910 MG
A mutagenic heterocyclic amine; mutagenic to S. typhimurium in the Ames test; induces tumor formation in the Zymbal gland, oral cavity, and colon in male and female rats, the skin of male rats, and the mammary gland in female rats at 0.03% in the diet