Learn More
Cayman Chemical 2AmIno3 8dImethylImIdazo 10mg
SDP
Catalog No. 502911309
Encompass_Preferred
A chlorogenic acid with antioxidant activity scavenges DPPH and superoxide anion radicals in cell-free assays (IC50s 9 and 36 UM respectively)
Catalog No. 50-291-1309 Supplier Cayman Chemical Supplier No. 3481210 MG
A chlorogenic acid with antioxidant activity scavenges DPPH and superoxide anion radicals in cell-free assays (IC50s 9 and 36 UM respectively)