Learn More
Cayman Chemical 2AmIno3methylImIdazo4 5f 10mg
SDP
Catalog No. 502985845
Encompass_Preferred
A food-derived carcinogen; metabolized by CYP1A2 and conjugated by N-acetyltransferase or sulfotransferase to a metabolite that reacts with DNA to form adducts; induces formation of single-base substitutions and exon deletions and increases cell death in vitro; increases incidence of squamous cell hyperplasias in the stomach of p53 knockout mice; increases hepatocellular lesions in female mice regardless of p53 status
Catalog No. 50-298-5845 Supplier Cayman Chemical Supplier No. 2185310 MG
A food-derived carcinogen; metabolized by CYP1A2 and conjugated by N-acetyltransferase or sulfotransferase to a metabolite that reacts with DNA to form adducts; induces formation of single-base substitutions and exon deletions and increases cell death in vitro; increases incidence of squamous cell hyperplasias in the stomach of p53 knockout mice; increases hepatocellular lesions in female mice regardless of p53 status