Learn More
Cayman Chemical 2-Amino-3-methylimidazo[4,5-f]quinoline, Purity: ≥98%, CAS: 76180-96-6, 50 mg
SDP
Catalog No. 502986648
Encompass_Preferred
A food-derived carcinogen; metabolized by CYP1A2 and conjugated by N-acetyltransferase or sulfotransferase to a metabolite that reacts with DNA to form adducts; induces formation of single-base substitutions and exon deletions and increases cell death in vitro; increases incidence of squamous cell hyperplasias in the stomach of p53 knockout mice; increases hepatocellular lesions in female mice regardless of p53 status
Catalog No. 50-298-6648 Supplier Cayman Chemical Supplier No. 2185350 MG
A food-derived carcinogen; metabolized by CYP1A2 and conjugated by N-acetyltransferase or sulfotransferase to a metabolite that reacts with DNA to form adducts; induces formation of single-base substitutions and exon deletions and increases cell death in vitro; increases incidence of squamous cell hyperplasias in the stomach of p53 knockout mice; increases hepatocellular lesions in female mice regardless of p53 status