Learn More
Cayman Chemical (2-Amino-5-bromophenyl)(pyridin-2-yl)methanone, Purity: ≥98%, CAS: 1563-56-0, 5 mg
SDP
Catalog No. 502917459
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of benzodiazepines; a metabolite and an acidic degradation product of bromazepam; intended for research and forensic applications
Catalog No. 50-291-7459 Supplier Cayman Chemical Supplier No. 379405 MG
An analytical reference standard categorized as a precursor in the synthesis of benzodiazepines; a metabolite and an acidic degradation product of bromazepam; intended for research and forensic applications