Learn More
Cayman Chemical 2AmIno6 7dIhydroxy1 2 50mg
SDP
Catalog No. 502995151
Encompass_Preferred
A dopamine receptor agonist (EC50s = 3.5 and 0.65 μM in rat striatal and nucleus accumbens homogenates, respectively); stimulates production of cAMP in rat striatal homogenates, a process that is reduced by 70% in the presence of dopamine antagonist fluphenazine at a concentration of 1 μM; induces hyperlocomotion in rats when injected bilaterally at a dose of 100 nmol per side into the nucleus accumbens; increases cAMP production and glycoprotein secretion in the albumen gland of H. duryi (snail); reduces sucrose feeding in R. maderae (cockroach) nymphs
Catalog No. 50-299-5151 Supplier Cayman Chemical Supplier No. 2386650 MG
A dopamine receptor agonist (EC50s = 3.5 and 0.65 μM in rat striatal and nucleus accumbens homogenates, respectively); stimulates production of cAMP in rat striatal homogenates, a process that is reduced by 70% in the presence of dopamine antagonist fluphenazine at a concentration of 1 μM; induces hyperlocomotion in rats when injected bilaterally at a dose of 100 nmol per side into the nucleus accumbens; increases cAMP production and glycoprotein secretion in the albumen gland of H. duryi (snail); reduces sucrose feeding in R. maderae (cockroach) nymphs