Learn More
Cayman Chemical 2AmInopurIn rIbosIde5Ot 2.5mg
SDP
Catalog No. 502918726
Encompass_Preferred
An isomer of ATP; can serve as a phosphate donor for ATP-requiring enzymes; inhibits AC from A. nicotianae (Ki = 1.7 mM)
Catalog No. 50-291-8726 Supplier Cayman Chemical Supplier No. 384352.5 MG
An isomer of ATP; can serve as a phosphate donor for ATP-requiring enzymes; inhibits AC from A. nicotianae (Ki = 1.7 mM)