Learn More
Cayman Chemical 2ArachIdonyl Glycerold5 100ug
SDP
Catalog No. 502913099
Encompass_Preferred
An internal standard for the quantification of 2-AG by GC- or LC-MS
Catalog No. 50-291-3099 Supplier Cayman Chemical Supplier No. 362162100 UG
An internal standard for the quantification of 2-AG by GC- or LC-MS