Learn More
ACROBiosystems ACROBiosystems Human 2B4 / CD244 / SLAMF4 Protein, His Tag
Catalog No. 501957522
Encompass_Preferred
Recombinant Protein;100UG;Human 2B4, His Tag (2B4-H5224) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # NP_057466).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.1 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, His Tag (Cat. No. 2B4-H5224) at 5 μg/mL (100 μL/well) can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-195-7522 Supplier ACROBiosystems Supplier No. 2B4H5224100UG
Recombinant Protein;100UG;Human 2B4, His Tag (2B4-H5224) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # NP_057466).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.1 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, His Tag (Cat. No. 2B4-H5224) at 5 μg/mL (100 μL/well) can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0.078-1.25 μg/mL (QC tested).