Learn More
ACROBiosystems Recombinant Protein;Human 2B4 / CD244 / SLAMF4 Protein, Fc Tag;HEK293;1MG;2B4,CD244,2B4,SLAMF4,NKR2B4,NAIL,h2B4,SLAMF-4
Supplier:  ACROBiosystems 2B4H52551MG
Encompass_Preferred
Recombinant Protein;Human 2B4 / CD244 / SLAMF4 Protein, Fc Tag;1MG;Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # NP_057466).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.9 kDa. As a result of glycosylation, the protein migrates as 65-80 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD48, His Tag (Cat. No. BC1-H5226) at 5 μg/mL (100 μL/well) can bind Human 2B4, Fc Tag (Cat. No. 2B4-H5255) with a linear range of 0.313-5 μg/mL (QC tested).
Catalog No. 50-165-9084