Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human 2B4 / CD244 / SLAMF4 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;2B4,CD244,2B4,SLAMF4,NKR2B4,NAIL,h2B4,SLAMF-4
Supplier:  ACROBiosystems 2B4H82F0200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human 2B4 / CD244 / SLAMF4 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human 2B4, Fc Tag (2B4-H82F0) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # NP_057466.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.7 kDa. As a result of glycosylation, the protein migrates as 65-80 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD48, Fc tag at 10 μg/mL (100 μL/well) can bind Biotinylated Human 2B4, Fc Tag (Cat. No. 2B4-H82F0) with a linear range of 0.01-0.156 μg/mL (QC tested).
Catalog No. 50-165-8452