Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human 2B4 / CD244 / SLAMF4 Protein (recommended for biopanning), His Tag & Avi Tag (Avitag™);HEK293;200UG;2B4,CD244,2B4,SLAMF4,NKR2B4,NAIL,h2B4,SLAMF-4
Catalog No. 501658254
Encompass_Preferred
Recombinant Protein;Biotinylated Human 2B4 / CD244 / SLAMF4 Protein (recommended for biopanning), His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human 2B4, His Tag (2B4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # Q9BZW8-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.4 kDa. The protein migrates as 40-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8254 Supplier ACROBiosystems Supplier No. 2B4H82E9200UG
Recombinant Protein;Biotinylated Human 2B4 / CD244 / SLAMF4 Protein (recommended for biopanning), His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human 2B4, His Tag (2B4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # Q9BZW8-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.4 kDa. The protein migrates as 40-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.