Learn More
Medchemexpress LLC HY-125021 5mg , 2BAct CAS:2143542-28-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1250215MG
Encompass_Preferred
HY-125021 5mg 2BAct CAS: 2143542-28-1 2BAct is a highly selective, and orally active eIF2B (eukaryotic initiation factor 2B) activator with an EC50 of 33 nM. 2BAct prevents neurological defects caused by a chronic integrated stress response. 2BAct is able to penetrate the central nervous system (CNS). 2BAct displays improved solubility and pharmacokinetics relative to eIF2B activator ISRIB (HY-12495). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0641