Learn More
Cayman Chemical 2C-B-FLY (hydrochloride), Purity: ≥98%, CAS: 178557-21-6, 10 mg
SDP
Catalog No. 502951536
Encompass_Preferred
A dihydrodifuran analog of the Schedule I hallucinogen 2C-B, which acts as a partial agonist at the 5-HT2A serotonin receptor; intended for forensic and research purposes
Catalog No. 50-295-1536 Supplier Cayman Chemical Supplier No. 1147310 MG
A dihydrodifuran analog of the Schedule I hallucinogen 2C-B, which acts as a partial agonist at the 5-HT2A serotonin receptor; intended for forensic and research purposes