Learn More
Cayman Chemical 2C-TFMhydrochlorIde 10mg
SDP
Catalog No. 502970720
Encompass_Preferred
A 2C compound with a para-trifluoromethyl substitution. It is a potent agonist of the 5-HT receptor subtypes 5-HT2A and 5-HT2C (Kaff = 165 and 32 nM); intended for forensic and research purposes
Catalog No. 50-297-0720 Supplier Cayman Chemical Supplier No. 1447110 MG
A 2C compound with a para-trifluoromethyl substitution. It is a potent agonist of the 5-HT receptor subtypes 5-HT2A and 5-HT2C (Kaff = 165 and 32 nM); intended for forensic and research purposes