Learn More
Cole-Parmer Thermo Scientific Nunc 363401 CryoTube cryogenic tube with round bottom; 1.8 mL, 50/bag
Catalog No. NC9634538 Shop All Cole-Parmer Products
Encompass_Preferred
Presterilized by cobalt-60 radiation, these one-piece molded polypropylene vials withstand temperatures from -196 to 135°C. Starfooted vials lock into CryoTube vial holder 03755-71 for one-handed operation. Optional colored cap inserts simplify sample identification. Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64.
This item is not returnable. View return policy
Catalog No. NC9634538 Supplier Cole-Parmer Supplier No. 0375520
Presterilized by cobalt-60 radiation, these one-piece molded polypropylene vials withstand temperatures from -196 to 135°C. Starfooted vials lock into CryoTube vial holder 03755-71 for one-handed operation. Optional colored cap inserts simplify sample identification. Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64.