Learn More
Instech Dual channel Vascular Access Harness™ for rats with one additional injection port. With VAH6M injector. Sterile. For rat infusion and blood sampling, or use with VAHD115L loop connector for bile sampling.
SDP
Catalog No. 5081252
Encompass
The Instech VAHD115AB‑1P is a sterile dual channel Vascular Access Harness™ for rats, featuring one additional injection port to support expanded dosing or sampling workflows. Supplied with a VAH6M injector for aseptic port access, it enables simultaneous infusion and blood sampling, and may also be paired with the VAHD115L loop connector for bile sampling applications. Designed for use with 3Fr polyurethane catheters, it provides reliable, low‑dead‑volume external access during both tethered and intermittent studies.
Features
  • Sterile dual channel Vascular Access Harness™ with extra injection port
  • Includes VAH6M injector for clean manual port access
  • Supports rat infusion and blood sampling workflows
  • Compatible with VAHD115L loop connector for bile sampling
  • Designed for 3Fr polyurethane catheter systems
Catalog No. 50-812-52 Supplier Instech Supplier No. VAHD115AB1P
The Instech VAHD115AB‑1P is a sterile dual channel Vascular Access Harness™ for rats, featuring one additional injection port to support expanded dosing or sampling workflows. Supplied with a VAH6M injector for aseptic port access, it enables simultaneous infusion and blood sampling, and may also be paired with the VAHD115L loop connector for bile sampling applications. Designed for use with 3Fr polyurethane catheters, it provides reliable, low‑dead‑volume external access during both tethered and intermittent studies.
Features
  • Sterile dual channel Vascular Access Harness™ with extra injection port
  • Includes VAH6M injector for clean manual port access
  • Supports rat infusion and blood sampling workflows
  • Compatible with VAHD115L loop connector for bile sampling
  • Designed for 3Fr polyurethane catheter systems