Learn More
Instech Dual channel Vascular Access Harness™ for rats. With VAH6M injector. Sterile. For rat infusion and blood sampling, or use with VAHD115L loop connector for bile sampling.
SDP
Catalog No. 5081251
Encompass
The Instech VAHD115AB is a sterile dual channel Vascular Access Harness™ for rats, supplied with a VAH6M injector for aseptic manual access to each channel. It supports simultaneous infusion and blood sampling, and can also be paired with the VAHD115L loop connector for bile sampling workflows. Designed for use with 3Fr catheters, it provides reliable, low‑dead‑volume external access during tethered or intermittent studies.
Features
  • Sterile dual channel Vascular Access Harness™ for rats
  • Includes VAH6M injector for manual aseptic access
  • Supports infusion and blood sampling applications
  • Compatible with VAHD115L loop connector for bile sampling
  • Designed for use with 3Fr polyurethane catheters
Catalog No. 50-812-51 Supplier Instech Supplier No. VAHD115AB
The Instech VAHD115AB is a sterile dual channel Vascular Access Harness™ for rats, supplied with a VAH6M injector for aseptic manual access to each channel. It supports simultaneous infusion and blood sampling, and can also be paired with the VAHD115L loop connector for bile sampling workflows. Designed for use with 3Fr catheters, it provides reliable, low‑dead‑volume external access during tethered or intermittent studies.
Features
  • Sterile dual channel Vascular Access Harness™ for rats
  • Includes VAH6M injector for manual aseptic access
  • Supports infusion and blood sampling applications
  • Compatible with VAHD115L loop connector for bile sampling
  • Designed for use with 3Fr polyurethane catheters