Learn More
Instech Dual channel Vascular Access Harness™ for rats with two additional injection ports. With VAH6M injector. Sterile. For rat infusion and blood sampling, or use with VAHD115L loop connector for bile sampling.
SDP
Catalog No. 5081253
Encompass
The Instech VAHD115AB‑2P is a sterile dual channel Vascular Access Harness™ for rats equipped with two additional injection ports to support expanded dosing and sampling workflows. Supplied with a VAH6M injector for aseptic manual access, it enables simultaneous infusion and blood sampling and may also be paired with the VAHD115L loop connector for bile sampling applications. Designed for use with 3Fr polyurethane catheters, it provides reliable, low‑dead‑volume access during tethered or intermittent studies.
Features
  • Sterile dual channel Vascular Access Harness™ with two extra injection ports
  • Includes VAH6M injector for clean manual port access
  • Supports rat infusion and blood sampling
  • Compatible with VAHD115L loop connector for bile sampling
  • Designed for 3Fr polyurethane catheter systems
Catalog No. 50-812-53 Supplier Instech Supplier No. VAHD115AB2P
The Instech VAHD115AB‑2P is a sterile dual channel Vascular Access Harness™ for rats equipped with two additional injection ports to support expanded dosing and sampling workflows. Supplied with a VAH6M injector for aseptic manual access, it enables simultaneous infusion and blood sampling and may also be paired with the VAHD115L loop connector for bile sampling applications. Designed for use with 3Fr polyurethane catheters, it provides reliable, low‑dead‑volume access during tethered or intermittent studies.
Features
  • Sterile dual channel Vascular Access Harness™ with two extra injection ports
  • Includes VAH6M injector for clean manual port access
  • Supports rat infusion and blood sampling
  • Compatible with VAHD115L loop connector for bile sampling
  • Designed for 3Fr polyurethane catheter systems