Learn More
Instech Two channel mouse Vascular Access Harness tether assembly, 7in, sterile.
SDP
Catalog No. 50890179
Encompass
The Instech VABM2T/25 is a two‑channel mouse VAB™ tether designed for simultaneous infusion and sampling when paired with a VABM2B‑series button. It includes two 10in (25cm) PU tubing lines—one clear and one blue‑stripe—each sized .017x.037in (0.43x0.94mm), a 7in (18cm) PS80 spring, and a 2‑channel 25ga connector. Red‑dot alignment ensures correct pairing with the red and white ports. Each channel has a 40µL dead volume, supporting low‑loss micro‑sampling workflows.
Features
  • 2‑channel mouse VAB™ tether
  • 10in (25cm) PU tubing, clear + blue‑stripe
  • .017x.037in (0.43x0.94mm) tubing size
  • 7in (18cm) PS80 spring
  • 25ga dual‑channel connector
  • 40µL dead volume per channel
Catalog No. 50-890-179 Supplier Instech Supplier No. VABM2T/25
The Instech VABM2T/25 is a two‑channel mouse VAB™ tether designed for simultaneous infusion and sampling when paired with a VABM2B‑series button. It includes two 10in (25cm) PU tubing lines—one clear and one blue‑stripe—each sized .017x.037in (0.43x0.94mm), a 7in (18cm) PS80 spring, and a 2‑channel 25ga connector. Red‑dot alignment ensures correct pairing with the red and white ports. Each channel has a 40µL dead volume, supporting low‑loss micro‑sampling workflows.
Features
  • 2‑channel mouse VAB™ tether
  • 10in (25cm) PU tubing, clear + blue‑stripe
  • .017x.037in (0.43x0.94mm) tubing size
  • 7in (18cm) PS80 spring
  • 25ga dual‑channel connector
  • 40µL dead volume per channel