Learn More
Cayman Chemical 2-Chloro-4-nitrophenyl α-D-maltotrioside, Purity: ≥95%, CAS: 118291-90-0, 1 g
SDP
Catalog No. 502950492
Encompass_Preferred
A colorimetric α-amylase substrate; upon hydrolysis by α-amylases, 2-chloro-4-nitrophenol is released which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity; has been used to characterize the activity of human pancreatic or salivary amylase
Catalog No. 50-295-0492 Supplier Cayman Chemical Supplier No. 344591 G
A colorimetric α-amylase substrate; upon hydrolysis by α-amylases, 2-chloro-4-nitrophenol is released which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity; has been used to characterize the activity of human pancreatic or salivary amylase