Learn More
Cayman Chemical 2-Chloro-4-nitrophenyl-α-D-glucopyranoside, Purity: ≥98%, CAS: 119047-14-2, 100 mg
SDP
Catalog No. 502965372
Encompass_Preferred
A conjugate of CNP and α-D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and α-D-glucose
Catalog No. 50-296-5372 Supplier Cayman Chemical Supplier No. 16527100 MG
A conjugate of CNP and α-D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and α-D-glucose