Learn More
Cayman Chemical 2-Chloro-4-nitrophenyl-α-D-glucopyranoside, Purity: ≥98%, CAS: 119047-14-2, 50 mg
SDP
Catalog No. 502965329
Encompass_Preferred
A conjugate of CNP and α-D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and α-D-glucose
Catalog No. 50-296-5329 Supplier Cayman Chemical Supplier No. 1652750 MG
A conjugate of CNP and α-D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and α-D-glucose