Learn More
Cayman Chemical 2-Chloroadenosine-5'-O-diphosphate (sodium salt), Purity: ≥95%, CAS: 82927-78-4, 2.3 mg
SDP
Catalog No. 502916679
Encompass_Preferred
A derivative of ADP; induces aggregation of, and inhibits PGE2-induced production of cAMP in, isolated human platelet-rich plasma; induces relaxation of precontracted isolated guinea pig taenia coli strips (pD2 = 6.74); decreases arterial blood pressure in rats; inhibits the ATPase activity of mortalin (apparent Ki = 45.05 µM for the human enzyme)
Catalog No. 50-291-6679 Supplier Cayman Chemical Supplier No. 384472.3 MG
A derivative of ADP; induces aggregation of, and inhibits PGE2-induced production of cAMP in, isolated human platelet-rich plasma; induces relaxation of precontracted isolated guinea pig taenia coli strips (pD2 = 6.74); decreases arterial blood pressure in rats; inhibits the ATPase activity of mortalin (apparent Ki = 45.05 µM for the human enzyme)