Learn More
Cayman Chemical 2-Chloro-N-methyl-N-(4-nitrophenyl)acetamide, Purity: ≥98%, CAS: 2653-16-9, 100 mg
SDP
Catalog No. 502909876
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of MELK inhibitors with anticancer activity
Catalog No. 50-290-9876 Supplier Cayman Chemical Supplier No. 35536100 MG
A synthetic intermediate; has been used in the synthesis of MELK inhibitors with anticancer activity