Learn More
AdipoGen 1-(2,5-Dichloro-3,6-dimethylphenyl ethanone (1 g)
SDP
Supplier:  AdipoGen CDXD0269G001
Encompass_Preferred
Chemical. CAS: 164165-77-9. Formula: C10H10Cl2O. MW: 217.1. Synthetic. Building block and intermediate for synthesis. Used for the synthesis of fluorescent dyes, such as chlorofluoresceincarboxylic acids.
Catalog No. 50-205-4375