Learn More
Cayman Chemical 2-Chloroadenosine 5-triphosphate (sodium salt), Purity: ≥95%, CAS: 301334-89-4, 2.7 mg
SDP
Catalog No. 502910852
Encompass_Preferred
An adenine nucleotide and ATP analog; antagonist of the purinergic P2Y1 receptor (Ki = 2.3 µM in Jurkat cells expressing the human receptor); an agonist of purinergic P2X receptors, inducing inward currents in HEK293 cells expressing the human bladder smooth muscle or rat PC12 forms of the receptor (EC50s = 0.5 and 2.5 µM, respectively); induces relaxation of precontracted isolated guinea pig taenia caeci strips in a concentration-dependent manner; has been used to study the substrate specificity of cyclic nucleotide-dependent protein kinases
Catalog No. 50-291-0852 Supplier Cayman Chemical Supplier No. 349782.7 MG
An adenine nucleotide and ATP analog; antagonist of the purinergic P2Y1 receptor (Ki = 2.3 µM in Jurkat cells expressing the human receptor); an agonist of purinergic P2X receptors, inducing inward currents in HEK293 cells expressing the human bladder smooth muscle or rat PC12 forms of the receptor (EC50s = 0.5 and 2.5 µM, respectively); induces relaxation of precontracted isolated guinea pig taenia caeci strips in a concentration-dependent manner; has been used to study the substrate specificity of cyclic nucleotide-dependent protein kinases