Learn More
Cayman Chemical 2-chloro-N6-Cyclopentyladenosine (hydrate), Purity: ≥98%, CAS: 1217443-91-8, 50 mg
SDP
Catalog No. 502939154
Encompass_Preferred
An adenosine receptor agonist; binds selectively to A1 over A2A and A3 receptors (Kis = 0.83, 2,300 and 42 nM, respectively, for the human receptors expressed in CHO cells; decreases heart rate in isolated rat atria (EC50 = 8.2 nM) but does not affect vasodilation in bovine coronary arteries; inhibits isoniazid- and pentylenetetrazole-induced convulsions in mice at 8.3 μmol/kg
Catalog No. 50-293-9154 Supplier Cayman Chemical Supplier No. 2873150 MG
An adenosine receptor agonist; binds selectively to A1 over A2A and A3 receptors (Kis = 0.83, 2,300 and 42 nM, respectively, for the human receptors expressed in CHO cells; decreases heart rate in isolated rat atria (EC50 = 8.2 nM) but does not affect vasodilation in bovine coronary arteries; inhibits isoniazid- and pentylenetetrazole-induced convulsions in mice at 8.3 μmol/kg