Learn More
Cayman Chemical 2-chloro-N6-Cyclopentyladenosine, Purity: ≥98%, CAS: 37739-05-2, 50 mg
SDP
Catalog No. 502913079
Encompass_Preferred
An adenosine A1 receptor antagonist; selectively binds to adenosine A1 over adenosine A2 receptors (Kis = 0.4 and 3,900 nM, respectively); inhibits forskolin-induced adenylate cyclase activity in rat fat cell membranes (IC50 = 33 nM); increases the electroconvulsive threshold in a mouse model of MES-induced seizures at 0.25 and 0.5 mg/kg; reduces infarct size in a rabbit model of coronary artery occlusion-induced myocardial infarction at 0.125 mg/kg
Catalog No. 50-291-3079 Supplier Cayman Chemical Supplier No. 3647750 MG
An adenosine A1 receptor antagonist; selectively binds to adenosine A1 over adenosine A2 receptors (Kis = 0.4 and 3,900 nM, respectively); inhibits forskolin-induced adenylate cyclase activity in rat fat cell membranes (IC50 = 33 nM); increases the electroconvulsive threshold in a mouse model of MES-induced seizures at 0.25 and 0.5 mg/kg; reduces infarct size in a rabbit model of coronary artery occlusion-induced myocardial infarction at 0.125 mg/kg