Learn More
Cayman Chemical 2-Chlorophenyl Cyclopentyl Ketone, Purity: ≥95%, CAS: 6740-85-8, 500 mg
SDP
Catalog No. 502942233
Encompass_Preferred
A starting material in the synthesis of ketamine and norketamine
Catalog No. 50-294-2233 Supplier Cayman Chemical Supplier No. 39809500 MG
A starting material in the synthesis of ketamine and norketamine