Learn More
Chemglass Life Sciences 2D Bag, 2L, 3 Port, (1) 1/8" & (2) 3/8" C-Flex x 12" Length, Plugged with Pinch Clamps, Sterile
SDP
Catalog No. 5000083189
Encompass
CG Biosciences CLS-1230 3-Port single-use bags are configured with plugs and pinch clamps.These 2D bags assembled in our ISO Class VII clean rooms.Bags are manufactured from medical-grade Renolit 9101 multilayer film composed of a high-purity biocompatible polyethylene contact layer with an internal ethylene vinyl alcohol oxygen-barrier.Bags are supplied gamma sterilized.Each bag has a grommeted hole or slot for hanging or supporting. ..Chemglass can provide different bags sizes, custom bag configurations and manifold assemblies:..500mL, 5L, 10L, 20L 3-Port bags are available upon request.To get started please contact us at singleuse@cglifesciences. comwith a sketch, diagram, or verbal description of the design you are looking for with the quantity required and one of our Bioprocess specialists will assist you.
Catalog No. 50-000-83189 Supplier Chemglass Life Sciences Supplier No. CLS12302000
CG Biosciences CLS-1230 3-Port single-use bags are configured with plugs and pinch clamps.These 2D bags assembled in our ISO Class VII clean rooms.Bags are manufactured from medical-grade Renolit 9101 multilayer film composed of a high-purity biocompatible polyethylene contact layer with an internal ethylene vinyl alcohol oxygen-barrier.Bags are supplied gamma sterilized.Each bag has a grommeted hole or slot for hanging or supporting. ..Chemglass can provide different bags sizes, custom bag configurations and manifold assemblies:..500mL, 5L, 10L, 20L 3-Port bags are available upon request.To get started please contact us at singleuse@cglifesciences. comwith a sketch, diagram, or verbal description of the design you are looking for with the quantity required and one of our Bioprocess specialists will assist you.