Learn More
Chemglass Life Sciences 2D Bag, 500mL, 2 Port, 1/4" ID x 3/8" OD C-Flex x 12" Length, Plugged with Pinch Clamps, Sterile
SDP
Catalog No. 5000083108
Encompass
CG Biosciences CLS-1230 2-Port single-use bags are configured with plugs and pinch clamps.These 2D bags assembled in our ISO Class VII clean rooms.Bags are manufactured from medical-grade Renolit 9101 multilayer film composed of a high-purity biocompatible polyethylene contact layer with an internal ethylene vinyl alcohol oxygen-barrier.Bags are supplied gamma sterilized.Each bag has a grommeted hole or slot for hanging or supporting.
Catalog No. 50-000-83108 Supplier Chemglass Life Sciences Supplier No. CLS12300500
CG Biosciences CLS-1230 2-Port single-use bags are configured with plugs and pinch clamps.These 2D bags assembled in our ISO Class VII clean rooms.Bags are manufactured from medical-grade Renolit 9101 multilayer film composed of a high-purity biocompatible polyethylene contact layer with an internal ethylene vinyl alcohol oxygen-barrier.Bags are supplied gamma sterilized.Each bag has a grommeted hole or slot for hanging or supporting.